Merck
  • Local Action of Neprilysin Exacerbates Pressure Overload Induced Cardiac Remodeling.
Local Action of Neprilysin Exacerbates Pressure Overload Induced Cardiac Remodeling.
Hypertension (Dallas, Tex. : 1979) (2021-04-13)
Hitoshi Nakagawa, Takuya Kumazawa, Kenji Onoue, Yasuki Nakada, Tomoya Nakano, Satomi Ishihara, Naoto Minamino, Hiroshi Hosoda, Nobuhisa Iwata, Tomoya Ueda, Ayako Seno, Taku Nishida, Tsunenari Soeda, Satoshi Okayama, Makoto Watanabe, Rika Kawakami, Yoshihiko Saito
ABSTRACT
[Figure: see text].