Merck
  • Intranasal type I interferon treatment is beneficial only when administered before clinical signs onset in the SARS-CoV-2 hamster model.
Intranasal type I interferon treatment is beneficial only when administered before clinical signs onset in the SARS-CoV-2 hamster model.
PLoS pathogens (2021-08-10)
Pierre Bessière, Marine Wasniewski, Evelyne Picard-Meyer, Alexandre Servat, Thomas Figueroa, Charlotte Foret-Lucas, Amelia Coggon, Sandrine Lesellier, Frank Boué, Nathan Cebron, Blandine Gausserès, Catherine Trumel, Gilles Foucras, Francisco J Salguero, Elodie Monchatre-Leroy, Romain Volmer
ABSTRACT
Impaired type I interferons (IFNs) production or signaling have been associated with severe COVID-19, further promoting the evaluation of recombinant type I IFNs as therapeutics against SARS-CoV-2 infection. In the Syrian hamster model, we show that intranasal administration of IFN-α starting one day pre-infection or one day post-infection limited weight loss and decreased viral lung titers. By contrast, intranasal administration of IFN-α starting at the onset of symptoms three days post-infection had no impact on the clinical course of SARS-CoV-2 infection. Our results provide evidence that early type I IFN treatment is beneficial, while late interventions are ineffective, although not associated with signs of enhanced disease.