Merck
  • Influence of Cooking Methods on Onion Phenolic Compounds Bioaccessibility.
Influence of Cooking Methods on Onion Phenolic Compounds Bioaccessibility.
Foods (Basel, Switzerland) (2021-06-03)
Alice Cattivelli, Angela Conte, Serena Martini, Davide Tagliazucchi
ABSTRACT
The impact of domestic cooking (baking, boiling, frying and grilling) and in vitro digestion on the stability and release of phenolic compounds from yellow-skinned (YSO) and red-skinned onions (RSO) have been evaluated. The mass spectrometry identification pointed out flavonols as the most representative phenolic class, led by quercetin-derivatives. RSO contained almost the double amount of phenolic compounds respect to YSO (50.12 and 27.42 mg/100 g, respectively). Baking, grilling and primarily frying resulted in an increased amount of total phenolic compounds, especially quercetin-derivatives, in both the onion varieties. Some treatments promoted the degradation of quercetin-3-O-hexoside-4'-O-hexoside, the main compound present in both the onion varieties, leading to the occurrence of quercetin-4'-O-hexoside and protocatechuic acid-4-O-hexoside. After in vitro digestion, the bioaccessibility index for total phenolic compounds ranged between 42.6% and 65.5% in grilled and baked YSO, respectively, and between 39.8% and 80.2% in boiled and baked RSO, respectively. Baking contributed to the highest amount of bioaccessible phenolic compounds for both the onion varieties after in vitro digestion. An in-depth design of the cooking process may be of paramount importance in modulating the gastro-intestinal release of onion phenolic compounds.