Merck
  • Heparin and Arginine Based Plasmin Nanoformulation for Ischemic Stroke Therapy.
Heparin and Arginine Based Plasmin Nanoformulation for Ischemic Stroke Therapy.
International journal of molecular sciences (2021-11-14)
Ramsha Aamir, Cameron Fyffe, Netanel Korin, Daniel A Lawrence, Enming J Su, Mathumai Kanapathipillai
ABSTRACT
Ischemic stroke is the most common type of stroke and thrombolytic therapy is the only approved treatment. However, current thrombolytic therapy with tissue plasminogen activator (tPA) is often hampered by the increased risk of hemorrhage. Plasmin, a direct fibrinolytic, has a significantly superior hemostatic safety profile; however, if injected intravenously it becomes rapidly inactivated by anti-plasmin. Nanoformulations have been shown to increase drug stability and half-life and hence could be applied to increase the plasmin therapeutic efficacy. Here in this paper, we report a novel heparin and arginine-based plasmin nanoformulation that exhibits increased plasmin stability and efficacy. In vitro studies revealed significant plasmin stability in the presence of anti-plasmin and efficient fibrinolytic activity. In addition, these particles showed no significant toxicity or oxidative stress effects in human brain microvascular endothelial cells, and no significant blood brain barrier permeability. Further, in a mouse photothrombotic stroke model, plasmin nanoparticles exhibited significant efficacy in reducing stroke volume without overt intracerebral hemorrhage (ICH) compared to free plasmin treatment. The study shows the potential of a plasmin nanoformulation in ischemic stroke therapy.
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg