Merck
Elevated serotonin coordinates mammary metabolism in dairy cows.
Physiological reports (2021-04-10)
Meghan K Connelly, Samantha R Weaver, Jordan M Kuehnl, Hannah P Fricke, Marisa Klister, Laura Hernandez
ABSTRACT
Serotonin plays a diverse role in maternal and mammary metabolism. Recent research in the dairy cow has shown a relationship between serotonin and calcium, with increased serotonin concentrations improving calcium homeostasis in the peri-partum dairy cow. Therefore, the objective was to elucidate how administration of 5-hydroxy-l-tryptophan (5-HTP), the immediate precursor to serotonin, altered serotonin and calcium metabolism in lactating dairy cows. Twelve mid-late lactation multiparous cows were blocked by parity, production and days in milk and allocated to a daily intravenous infusion of (i) 1.5 mg/kg of 5-HTP (n = 6) or (ii) saline (n = 6) for 3 consecutive days. Milk samples were collected daily. Blood samples were collected before and after each infusion with mammary biopsies and blood samples collected at 48, 56, and 72 h relative to termination of first infusion. Infusion of 5-HTP increased (p = 0.001) circulating serotonin concentrations and decreased blood calcium via a transient hypocalcemia immediately after each infusion (p = 0.02). Treatment with 5-HTP increased milk calcium concentrations (p = 0.02) and calcium release-activated channel protein 1 (ORAI1) mRNA at 56 h and protein at 48 h relative to termination of first infusion (p = 0.008 and p = 0.09, respectively). Fifty-six hours from termination of the first infusion mRNA of parathyroid hormone-related protein and mammary serotonin content were increased relative to control (p = 0.03 and p = 0.05, respectively). These findings demonstrate the ability of 5-HTP infusion to increase circulating serotonin concentrations and alter endocrine and mammary autocrine/paracrine calcium and serotonin metabolism in the lactating dairy cow.