Merck
  • Protective Effects of Six Selected Dietary Compounds against Leptin-Induced Proliferation of Oestrogen Receptor Positive (MCF-7) Breast Cancer Cells.
Protective Effects of Six Selected Dietary Compounds against Leptin-Induced Proliferation of Oestrogen Receptor Positive (MCF-7) Breast Cancer Cells.
Medicines (Basel, Switzerland) (2017-09-21)
Meran Keshawa Ediriweera, Kamani Hemamala Tennekoon, Sameera Ranganath Samarakoon, Ira Thabrew, E Dilip de Silva
ABSTRACT
Obesity is considered as one of the risk factors for breast cancer. Leptin has been found to be involved in breast cancer progression. Therefore, novel approaches to antagonize biological effects of leptin are much needed. The objective of this study was to evaluate the protective effects of six dietary compounds (quercetin, curcumin, gallic acid, epigallocatechin gallate (EGCG), ascorbic acid and catechin) and assess the phosphorylation of extracellular signal-regulated kinase 1/2 (ERK1/2) in leptin-stimulated MCF-7 breast cancer cells in vitro. MCF-7 cells were exposed to leptin, leptin and compound and compound alone for 48 h. Cell viability was determined by 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide MTT and fluorometric assays after 48 h incubation. Phosphorylation of ERK1/2 was quantified by ELISA. Only quercetin, curcumin and EGCG showed significant protective effects against leptin-induced proliferation of MCF-7 cells. Increase in ERK1/2 phosphorylation in response to leptin was reduced by the addition of quercetin, curcumin and EGCG. Considering the high prevalence of obesity, this observation provides a rationale for use of curcumin, quercetin and EGCG as antagonists of leptin in the treatment of obese breast cancer patients.