Merck
  • The calcium/calmodulin-dependent phosphodiesterase PDE1C down-regulates glucose-induced insulin secretion.
The calcium/calmodulin-dependent phosphodiesterase PDE1C down-regulates glucose-induced insulin secretion.
The Journal of biological chemistry (1999-07-31)
P Han, J Werber, M Surana, N Fleischer, T Michaeli
ABSTRACT
To understand the role cAMP phosphodiesterases (PDEs) play in the regulation of insulin secretion, we analyzed cyclic nucleotide PDEs of a pancreatic beta-cell line and used family and isozyme-specific PDE inhibitors to identify the PDEs that counteract glucose-stimulated insulin secretion. We demonstrate the presence of soluble PDE1C, PDE4A and 4D, a cGMP-specific PDE, and of particulate PDE3, activities in betaTC3 insulinoma cells. Selective inhibition of PDE1C, but not of PDE4, augmented glucose-stimulated insulin secretion in a dose-dependent fashion thus demonstrating that PDE1C is the major PDE counteracting glucose-dependent insulin secretion from betaTC3 cells. In pancreatic islets, inhibition of both PDE1C and PDE3 augmented glucose-dependent insulin secretion. The PDE1C of betaTC3 cells is a novel isozyme possessing a K(m) of 0.47 microM for cAMP and 0.25 microM for cGMP. The PDE1C isozyme of betaTC3 cells is sensitive to 8-methoxymethyl isobutylmethylxanthine and zaprinast (IC(50) = 7.5 and 4.5 microM, respectively) and resistant to vinpocetine (IC(50) > 100 microM). Increased responsiveness of PDE1C activity to calcium/calmodulin is evident upon exposure of cells to glucose. Enhanced cAMP degradation by PDE1C, due to increases in its responsiveness to calcium/calmodulin and in intracellular calcium, constitutes a glucose-dependent feedback mechanism for the control of insulin secretion.