- An asymmetric mechanical code ciphers curvature-dependent proprioceptor activity.
An asymmetric mechanical code ciphers curvature-dependent proprioceptor activity.
Science advances (2021-09-18)
Ravi Das, Li-Chun Lin, Frederic Català-Castro, Nawaphat Malaiwong, Neus Sanfeliu-Cerdán, Montserrat Porta-de-la-Riva, Aleksandra Pidde, Michael Krieg
PMID34533987
ABSTRACT
[Figure: see text].