Merck
  • COVID-19 antibody screening with SARS-CoV-2 red cell kodecytes using routine serologic diagnostic platforms.
COVID-19 antibody screening with SARS-CoV-2 red cell kodecytes using routine serologic diagnostic platforms.
Transfusion (2021-02-17)
Radhika Nagappan, Willy A Flegel, Kshitij Srivastava, Eleanor C Williams, Ivan Ryzhov, Alexander Tuzikov, Oxana Galanina, Nadezhda Shilova, Gennady Sukhikh, Holly Perry, Nicolai V Bovin, Stephen M Henry
ABSTRACT
The Coronavirus disease 2019 (COVID-19) pandemic is having a major global impact, and the resultant response in the development of new diagnostics is unprecedented. The detection of antibodies against severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) has a role in managing the pandemic. We evaluated the feasibility of using SARS-CoV-2 peptide Kode Technology-modified red cells (C19-kodecytes) to develop an assay compatible with existing routine serologic platforms. A panel of eight unique red cells modified using Kode Technology function-spacer-lipid constructs and bearing short SARS-CoV-2 peptides was developed (C19-kodecyte assay). Kodecytes were tested against undiluted expected antibody-negative and -positive plasma samples in manual tube and three column agglutination technology (CAT) platforms. Parallel analysis with the same peptides in solid phase by enzyme immunoassays was performed. Evaluation samples included >120 expected negative blood donor samples and >140 COVID-19 convalescent plasma samples, with independent serologic analysis from two centers. Specificity (negative reaction rate against expected negative samples) in three different CAT platforms against novel C19-kodecytes was >91%, which correlated with published literature. Sensitivity (positive reaction rate against expected positive convalescent, PCR-confirmed samples) ranged from 82% to 97% compared to 77% with the Abbott Architect SARS-CoV-2 IgG assay. Manual tube serology was less sensitive than CAT. Enzyme immunoassay results with some Kode Technology constructs also had high sensitivity. C19-kodecytes are viable for use as serologic reagent red cells for the detection of SARS-CoV-2 antibody with routine blood antibody screening equipment.