- Peptide mapping of polypeptides separated by two-dimensional electrophoresis: protease digestion directly on the two-dimensional gel followed by electrophoresis in reverse direction.
Peptide mapping of polypeptides separated by two-dimensional electrophoresis: protease digestion directly on the two-dimensional gel followed by electrophoresis in reverse direction.
Electrophoresis (1988-07-01)
M Zivy, F Granier
PMID3069460
ABSTRACT
A method is described which allows to reveal simultaneously the proteolytic patterns of numerous polypeptides separated by two-dimensional electrophoresis. After two-dimensional electrophoresis, the gels were dipped successively in buffers for preequilibration, protease digestion, and reequilibration. They were then returned to the electrophoresis tank, and electrophoresis was continued for a short time. After silver staining, digestion products appeared, lined up behind the original polypeptide spots. The method allows proteolytic patterns of numerous polypeptides to be visualized simply and quickly. Among proteins of wheat leaves, 31 groups of related polypeptides were found according to the similarity of their proteolytic patterns.