Merck
  • Alterations in proteins of bone marrow extracellular matrix in undernourished mice.
Alterations in proteins of bone marrow extracellular matrix in undernourished mice.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas (2000-08-02)
C L Vituri, M Alvarez-Silva, A G Trentin, P Borelli
ABSTRACT
The objective of the present study was to determine the effect of protein malnutrition on the glycoprotein content of bone marrow extracellular matrix (ECM). Two-month-old male Swiss mice were submitted to protein malnutrition with a low-protein diet containing 4% casein as compared to 20% casein in the control diet. When the experimental group had attained a 20% loss of their original body weight, we extracted the ECM proteins from bone marrow with PBS buffer, and analyzed ECM samples by SDS-PAGE (7.5%) and ECL Western blotting. Quantitative differences were observed between control and experimental groups. Bone marrow ECM from undernourished mice had greater amounts of extractable fibronectin (1.6-fold increase) and laminin (4.8-fold increase) when compared to the control group. These results suggest an association between fluctuations in the composition of the hematopoietic microenvironment and altered hematopoiesis observed in undernourished mice.