Merck
  • Application of counter-selectable marker PIGA in engineering designer deletion cell lines and characterization of CRISPR deletion efficiency.
Application of counter-selectable marker PIGA in engineering designer deletion cell lines and characterization of CRISPR deletion efficiency.
Nucleic acids research (2021-02-17)
Donghui Li, Xiaoji Sun, Fangzhou Yu, Mary Ann Perle, David Araten, Jef D Boeke
ABSTRACT
The CRISPR/Cas9 system is a technology for genome engineering, which has been applied to indel mutations in genes as well as targeted gene deletion and replacement. Here, we describe paired gRNA deletions along the PIGA locus on the human X chromosome ranging from 17 kb to 2 Mb. We found no compelling linear correlation between deletion size and the deletion efficiency, and there is no substantial impact of topologically associating domains on deletion frequency. Using this precise deletion technique, we have engineered a series of designer deletion cell lines, including one with deletions of two X-chromosomal counterselectable (negative selection) markers, PIGA and HPRT1, and additional cell lines bearing each individual deletion. PIGA encodes a component of the glycosylphosphatidylinositol (GPI) anchor biosynthetic apparatus. The PIGA gene counterselectable marker has unique features, including existing single cell level assays for both function and loss of function of PIGA and the existence of a potent counterselectable agent, proaerolysin, which we use routinely for selection against cells expressing PIGA. These designer cell lines may serve as a general platform with multiple selection markers and may be particularly useful for large scale genome engineering projects such as Genome Project-Write (GP-write).