Merck
  • Characterization of a newly established schwannoma cell line from a sporadic vestibular schwannoma patient.
Characterization of a newly established schwannoma cell line from a sporadic vestibular schwannoma patient.
American journal of translational research (2021-09-21)
Lu Xue, Weiwei He, Zhigang Wang, Hongsai Chen, Zhaoyan Wang, Hao Wu
ABSTRACT
A stable, human sporadic vestibular schwannoma cell line is not currently available. By using a lentivirus-mediated transfection from a 41-year-old sporadic vestibular schwannoma patient, primary schwannoma cells were obtained, cultured and immortalized using the hHERT gene. The NF2 gene of the resulting JEI-001 cell line contains a specific Exon 5 mutation. The schwannoma cell origin of this cell line was confirmed using STR techniques and immunocytochemistry. A comparison between the primary tumor tissue and JEI-001 revealed a common mutation of the NF2 gene, which indicated that the JEI-001 cell line had retained most of its original tumor characteristics. The JEI-001 cell line was found to be non-tumorigenic in nude mice, but certain growth features had been altered, resulting in changes such as independence from the Schwann cell growth factors and a higher proliferation rate. This was the first known study to establish cell lines immortalized from human sporadic vestibular schwannoma that had different characteristics from that of HEI-193. This is a novel model system that can be used for the study of NF2 gene functions, in order to elaborate on the biological features of sporadic vestibular schwannoma, even including familial NF2 tumors, and to further explore the molecular pathogenesis and develop new adjuvant therapies.