Merck
  • Immunocompromised Cas9 transgenic mice for rapid in vivo assessment of host factors involved in highly pathogenic virus infection.
Immunocompromised Cas9 transgenic mice for rapid in vivo assessment of host factors involved in highly pathogenic virus infection.
Molecular therapy. Methods & clinical development (2021-11-04)
Nicole Collette, Pragyesh Dhungel, Sean J Lund, Jennifer L Schwedler, Edwin A Saada, Yooli K Light, Anupama Sinha, Joseph S Schoeniger, Oscar A Negrete
ABSTRACT
Targeting host factors for anti-viral development offers several potential advantages over traditional countermeasures that include broad-spectrum activity and prevention of resistance. Characterization of host factors in animal models provides strong evidence of their involvement in disease pathogenesis, but the feasibility of performing high-throughput in vivo analyses on lists of genes is problematic. To begin addressing the challenges of screening candidate host factors in vivo, we combined advances in CRISPR-Cas9 genome editing with an immunocompromised mouse model used to study highly pathogenic viruses. Transgenic mice harboring a constitutively expressed Cas9 allele (Cas9 tg/tg ) with or without knockout of type I interferon receptors served to optimize in vivo delivery of CRISPR single-guide RNA (sgRNA) using Invivofectamine 3.0, a simple and easy-to-use lipid nanoparticle reagent. Invivofectamine 3.0-mediated liver-specific editing to remove activity of the critical Ebola virus host factor Niemann-Pick disease type C1 in an average of 74% of liver cells protected immunocompromised Cas9 tg/tg mice from lethal surrogate Ebola virus infection. We envision that immunocompromised Cas9 tg/tg mice combined with straightforward sgRNA in vivo delivery will enable efficient host factor loss-of-function screening in the liver and other organs to rapidly study their effects on viral pathogenesis and help initiate development of broad-spectrum, host-directed therapies against emerging pathogens.