Merck
Structure and mechanism of alkaline phosphatase.
Annual review of biophysics and biomolecular structure (1992-01-01)
J E Coleman
ABSTRACT
Alkaline phosphatase was the first zinc enzyme to be discovered in which three closely spaced metal ions (two Zn ions and one Mg ion) are present at the active center. Zn ions at all three sites also produce a maximally active enzyme. These metal ions have center-to-center distances of 3.9 A (Zn1-Zn2), 4.9 A (Zn2-Mg3), and 7.1 A (Zn1-Mg3). Despite the close packing of these metal centers, only one bridging ligand, the carboxyl of Asp51, bridges Zn2 and Mg3. A crystal structure at 2.0-A resolution of the noncovalent phosphate complex, E.P, formed with the active center shows that two phosphate oxygens form a phosphate bridge between Zn1 and Zn2, while the two other phosphate oxygens form hydrogen bonds with the guanidium group of Arg166. This places Ser102, the residue known to be phosphorylated during phosphate hydrolysis, in the required apical position to initiate a nucleophilic attack on the phosphorous. Extrapolation of the E.P structure to the enzyme-substrate complex, E.ROPO4(2-), leads to the conclusion that Zn1 must coordinate the ester oxygen, thus activating the leaving group in the phosphorylation of Ser102. Likewise, Zn2 appears to coordinate the ester oxygen of the seryl phosphate and activate the leaving group during the hydrolysis of the phosphoseryl intermediate. Both of these findings suggest that there may be a significant dissociative character to each of the two displacements at phosphorous catalyzed by alkaline phosphatase. A water molecule (or hydroxide) coordinated to Zn1 following formation of the phosphoseryl intermediate appears to be the nucleophile in the second step of the mechanism. Dissociation of the product phosphate from the E.P intermediate is the slowest, 35 s-1, and therefore the rate-limiting, step of the mechanism at alkaline pH. Since the determination of the initial crystal structure of alkaline phosphatase, two other crystal structures of enzymes involved in phosphate ester hydrolysis have been completed that show a triad of closely spaced zinc ions present at their active centers. These enzymes are phospholipase C from Bacillus cereus (structure at 1.5-A resolution) (43) and P1 nuclease from Penicillium citrinum (structure at 2.8-A resolution) (74). Both enzymes hydrolyze phosphodiesters. Substrates for phospholipase C are phosphatidylinositol and phosphatidylcholine, while P1 nuclease is an endonuclease hydrolyzing single stranded ribo- and deoxyribonucleotides. P1 nuclease also has activity as a phosphomonoesterase against 3'-terminal phosphates of nucleotides. The Zn ions in both enzymes form almost identical trinuclear sites.(ABSTRACT TRUNCATED AT 400 WORDS)