Merck
  • Retinoid-resistant estrogen receptor-negative human breast carcinoma cells transfected with retinoic acid receptor-alpha acquire sensitivity to growth inhibition by retinoids.
Retinoid-resistant estrogen receptor-negative human breast carcinoma cells transfected with retinoic acid receptor-alpha acquire sensitivity to growth inhibition by retinoids.
The Journal of biological chemistry (1994-08-26)
M S Sheikh, Z M Shao, X S Li, M Dawson, A M Jetten, S Wu, B A Conley, M Garcia, H Rochefort, J A Fontana
ABSTRACT
Retinoids mediate their actions via RARs (retinoic acid receptors) and RXRs (retinoid X receptors). Each class of these nuclear retinoid receptors is further subdivided into three species, namely alpha, beta, and gamma. Recent studies demonstrate that estrogen receptor (ER)-positive human breast carcinoma (HBC) cell lines and tumor samples exhibit significantly higher levels of RAR alpha than their ER-negative counterparts. ER-positive HBC cell lines are sensitive to, and ER-negative cell lines are resistant to, growth inhibitory effects of retinoic acid (RA). We previously demonstrated that the expression of functional ERs in an established ER-negative cell line resulted in higher levels of RAR alpha and sensitivity to growth inhibition by RA. To further investigate the major role of RAR alpha in retinoid-mediated inhibition of growth, we transfected RAR alpha cDNA in two RA-resistant ER-negative HBC cell lines. Analyses of different clonal populations of RAR alpha transfectants from each cell line revealed growth inhibition by retinoids. Utilizing RAR- and RXR-class selective retinoids, we further demonstrated that only the RAR alpha-selective retinoids mediated the growth inhibition in these cells, while the RXR-selective retinoids were biologically inert. We thus provide evidence that the molecular mechanisms of retinoid inhibition of HBC proliferation predominantly involve RAR alpha.