Merck
  • Atherosclerosis Regression and Cholesterol Efflux in Hypertriglyceridemic Mice.
Atherosclerosis Regression and Cholesterol Efflux in Hypertriglyceridemic Mice.
Circulation research (2021-02-04)
Tatjana Josefs, Debapriya Basu, Tomas Vaisar, Britt Arets, Jenny E Kanter, Lesley-Ann Huggins, Yunying Hu, Jianhua Liu, Noemie Clouet-Foraison, Jay W Heinecke, Karin E Bornfeldt, Ira J Goldberg, Edward A Fisher
ABSTRACT
[Figure: see text].