Merck
  • Increase of 5-HT levels is induced both in mouse brain and HEK-293 cells following their exposure to a non-viral tryptophan hydroxylase construct.
Increase of 5-HT levels is induced both in mouse brain and HEK-293 cells following their exposure to a non-viral tryptophan hydroxylase construct.
Translational psychiatry (2021-10-10)
Emiliano Tesoro-Cruz, Leticia Manuel-Apolinar, Norma Oviedo, Sandra Orozco-Suárez, Minerva Crespo Ramírez, Vilma Carolina Bekker-Méndez, M Magdalena Aguirre-García, Sandra Angélica Rojas-Osornio, Vladimir Paredes-Cervantes, Miguel Pérez de la Mora
ABSTRACT
Tryptophan hydroxylase type 2 (Tph2) is the rate-limiting enzyme for serotonin (5-HT) biosynthesis in the brain. Dysfunctional Tph2 alters 5-HT biosynthesis, leading to a deficiency of 5-HT, which could have repercussions on human behavior. In the last decade, several studies have associated polymorphisms of the TPH2 gene with suicidal behavior. Additionally, a 5-HT deficiency has been implicated in various psychiatric pathologies, including alcoholism, impulsive behavior, anxiety, and depression. Therefore, the TPH2 gene could be an ideal target for analyzing the effects of a 5-HT deficiency on brain function. The aim of this study was to use the construct pIRES-hrGFP-1a-Tph2-FLAG to treat CD1-male mice and to transfect HEK-293-cells and then to evaluate whether this treatment increases 5-HT production. 5-HT levels were enhanced 48 h post-transfection, in HEK-293 cells. Three days after the ocular administration of pIRES-hrGFP-1a-Tph2-FLAG to mice, putative 5-HT production was significantly higher than in the control in both hypothalamus and amygdala, but not in the brainstem. Further research will be needed on the possible application of this treatment for psychiatric diseases involving a Tph2 dysfunction or serotonin deficiency.