- Bispecific antibodies targeting distinct regions of the spike protein potently neutralize SARS-CoV-2 variants of concern.
Bispecific antibodies targeting distinct regions of the spike protein potently neutralize SARS-CoV-2 variants of concern.
Science translational medicine (2021-09-15)
Hyeseon Cho, Kristina Kay Gonzales-Wartz, Deli Huang, Meng Yuan, Mary Peterson, Janie Liang, Nathan Beutler, Jonathan L Torres, Yu Cong, Elena Postnikova, Sandhya Bangaru, Chloe Adrienna Talana, Wei Shi, Eun Sung Yang, Yi Zhang, Kwanyee Leung, Lingshu Wang, Linghang Peng, Jeff Skinner, Shanping Li, Nicholas C Wu, Hejun Liu, Cherrelle Dacon, Thomas Moyer, Melanie Cohen, Ming Zhao, Frances Eun-Hyung Lee, Rona S Weinberg, Iyadh Douagi, Robin Gross, Connie Schmaljohn, Amarendra Pegu, John R Mascola, Michael Holbrook, David Nemazee, Thomas F Rogers, Andrew B Ward, Ian A Wilson, Peter D Crompton, Joshua Tan
PMID34519517
ABSTRACT
[Figure: see text].