Merck
  • Camptothecin-Induced Replication Stress Affects DNA Replication Profiling by E/L Repli-Seq.
Camptothecin-Induced Replication Stress Affects DNA Replication Profiling by E/L Repli-Seq.
Cytogenetic and genome research (2021-11-25)
Takuya Hayakawa, Rino Suzuki, Kazuhiro Kagotani, Katsuzumi Okumura, Shin-Ichiro Takebayashi
ABSTRACT
E/L Repli-seq is a powerful tool for detecting cell type-specific replication landscapes in mammalian cells, but its potential to monitor DNA replication under replication stress awaits better understanding. Here, we used E/L Repli-seq to examine the temporal order of DNA replication in human retinal pigment epithelium cells treated with the topoisomerase I inhibitor camptothecin. We found that the replication profiles by E/L Repli-seq exhibit characteristic patterns after replication-stress induction, including the loss of specific initiation zones within individual early replication timing domains. We also observed global disappearance of the replication timing domain structures in the profiles, which can be explained by checkpoint-dependent suppression of replication initiation. Thus, our results demonstrate the effectiveness of E/L Repli-seq at identifying cells with replication-stress-induced altered DNA replication programs.