Merck
  • An infectivity-enhancing site on the SARS-CoV-2 spike protein targeted by antibodies.
An infectivity-enhancing site on the SARS-CoV-2 spike protein targeted by antibodies.
Cell (2021-06-18)
Yafei Liu, Wai Tuck Soh, Jun-Ichi Kishikawa, Mika Hirose, Emi E Nakayama, Songling Li, Miwa Sasai, Tatsuya Suzuki, Asa Tada, Akemi Arakawa, Sumiko Matsuoka, Kanako Akamatsu, Makoto Matsuda, Chikako Ono, Shiho Torii, Kazuki Kishida, Hui Jin, Wataru Nakai, Noriko Arase, Atsushi Nakagawa, Maki Matsumoto, Yukoh Nakazaki, Yasuhiro Shindo, Masako Kohyama, Keisuke Tomii, Koichiro Ohmura, Shiro Ohshima, Toru Okamoto, Masahiro Yamamoto, Hironori Nakagami, Yoshiharu Matsuura, Atsushi Nakagawa, Takayuki Kato, Masato Okada, Daron M Standley, Tatsuo Shioda, Hisashi Arase
ABSTRACT
Antibodies against the receptor-binding domain (RBD) of the SARS-CoV-2 spike protein prevent SARS-CoV-2 infection. However, the effects of antibodies against other spike protein domains are largely unknown. Here, we screened a series of anti-spike monoclonal antibodies from coronavirus disease 2019 (COVID-19) patients and found that some of antibodies against the N-terminal domain (NTD) induced the open conformation of RBD and thus enhanced the binding capacity of the spike protein to ACE2 and infectivity of SARS-CoV-2. Mutational analysis revealed that all of the infectivity-enhancing antibodies recognized a specific site on the NTD. Structural analysis demonstrated that all infectivity-enhancing antibodies bound to NTD in a similar manner. The antibodies against this infectivity-enhancing site were detected at high levels in severe patients. Moreover, we identified antibodies against the infectivity-enhancing site in uninfected donors, albeit at a lower frequency. These findings demonstrate that not only neutralizing antibodies but also enhancing antibodies are produced during SARS-CoV-2 infection.