Merck
  • Chemotherapy regimens induce inhibitory immune checkpoint protein expression on stem-like and senescent-like oesophageal adenocarcinoma cells.
Chemotherapy regimens induce inhibitory immune checkpoint protein expression on stem-like and senescent-like oesophageal adenocarcinoma cells.
Translational oncology (2021-03-26)
Maria Davern, Noel E Donlon, Andrew Sheppard, Fiona O' Connell, Conall Hayes, Anshul Bhardwaj, Emma Foley, Dermot O' Toole, Niamh Lynam-Lennon, Narayanasamy Ravi, John V Reynolds, Stephen G Maher, Joanne Lysaght
ABSTRACT
Use of immune checkpoint inhibitors (ICIs) with chemotherapy to enhance responses in oesophageal adenocarcinoma (OAC) is an attractive approach. We identified subpopulations of OAC cells expressing inhibitory immune checkpoint (IC) ligands (PD-L1, PD-L2 and CD160) and receptors (PD-1, TIGIT, TIM-3, LAG-3 and A2aR) in vitro and in ex vivo biopsies. Combination chemotherapy regimens FLOT and CROSS promote a more immune-resistant phenotype through upregulation of IC ligands and receptors on OAC cells in vitro. Importantly, this study investigated if OAC cells, enriched for ICs exhibited a more stem-like and senescent-like phentoype. FLOT preferentially upregulates PD-L1 on a stem-like OAC cell phenotype, defined by ALDH activity. Expression of senescence-associated β-galactosidase is induced in a subpopulation of OAC cells following FLOT and CROSS chemotherapy treatment, along with enhanced expression of TIM-3 and A2aR ICs. Blockade of PD-1 signalling in OAC cells induced apoptosis and enhanced FLOT and CROSS chemotherapy toxicity in vitro. Upregulation of ICs on OAC cells following chemotherapy may represent potential mechanisms of chemo-immune resistance. Combination ICIs may be required to enhance the efficacy of chemotherapy and immunotherapy in OAC patients.