Merck
  • Kinetic mechanism of L-α-glycerophosphate oxidase from Mycoplasma pneumoniae.
Kinetic mechanism of L-α-glycerophosphate oxidase from Mycoplasma pneumoniae.
The FEBS journal (2015-02-26)
Somchart Maenpuen, Pratchaya Watthaisong, Pacharee Supon, Jeerus Sucharitakul, Derek Parsonage, P Andrew Karplus, Al Claiborne, Pimchai Chaiyen
ABSTRACT
L-α-glycerophosphate oxidase is an FAD-dependent enzyme that catalyzes the oxidation of L-α-glycerophosphate (Glp) by molecular oxygen to generate dihydroxyacetone phosphate (DHAP) and hydrogen peroxide (H2O2). The catalytic properties of recombinant His6-GlpO from Mycoplasma pneumoniae (His6-MpGlpO) were investigated through transient and steady-state kinetics and ligand binding studies. The results indicate that the reaction mechanism of His6-MpGlpO follows a ping-pong model. Double-mixing mode stopped-flow experiments show that, after flavin-mediated substrate oxidation, DHAP leaves rapidly prior to the oxygen reaction. The values determined for the individual rate constants and kcat (4.2 s(-1) at 4 °C), in addition to the finding that H2 O2 binds to the oxidized enzyme, suggest that H2O2 release is the rate-limiting step for the overall reaction. The results indicate that His6 -MpGlpO contains mixed populations of fast- and slow-reacting species. It is predominantly the fast-reacting species that participates in turnover. In contrast to other GlpO enzymes previously described, His6-MpGlpO is able to catalyze the reverse reaction of reduced enzyme and DHAP. This result may be explained by the standard reduction potential value of His6-MpGlpO (-167 ± 1 mV), which is lower than those of GlpO from other species. We found that D,L-glyceraldehyde 3-phosphate (GAP) may be used as a substrate in the His6-MpGlpO reaction, although it exhibited an approximately 100-fold lower kcat value in comparison with the reaction of Glp. These results also imply involvement of GlpO in glycolysis, as well as in lipid and glycerol metabolism. The kinetic models and distinctive properties of His6-MpGlpO reported here should be useful for future drug development against Mycoplasma pneumoniae infection.