Merck
  • Tumor imaging using P866, a high-relaxivity gadolinium chelate designed for folate receptor targeting.
Tumor imaging using P866, a high-relaxivity gadolinium chelate designed for folate receptor targeting.
Magnetic resonance in medicine (2008-11-26)
Claire Corot, Philippe Robert, Eric Lancelot, Philippe Prigent, Sébastien Ballet, Irène Guilbert, Jean-Sébastien Raynaud, Isabelle Raynal, Marc Port
ABSTRACT
The objective of this study was to evaluate the potential of a high-relaxivity macromolecular gadolinium (Gd) chelate to target folate receptors (FRs). P866 is a dimeric high-relaxivity Gd chelate coupled to a folate moiety. Binding affinity, in vivo biodistribution studies in KB tumor-bearing mice at 1, 4, and 24 h, and dynamic contrast-enhanced (DCE)-MRI (2.35 T) over 4 h were assessed. Binding and internalization of P866 through the FR was demonstrated. Due to the high molecular volume of P866, the binding affinity compared to free FA was decreased (K(D) = 59.3 +/- 1.8 nM and 5.9 +/- 0.2 nM, respectively). Tumor/muscle (T/M) uptake was 5.4 +/- 1.0, 4 h after injection of 15 micromol/kg. Competition with free FA was less effective when the dose was increased due to a saturation of FR. At a dose of 5 micromol/kg, a 70% difference in signal enhancement was observed between P866 and the nonspecific reference compound, thus demonstrating the specificity of FR targeting. While this high-relaxivity folate-Gd chelate has demonstrated its potential capacity to target in vivo FR on tumors, the sensitivity is probably limited to a certain extent by the saturation of the FR and by the decrease in the apparent relaxivity of the internalized part of P866 in the tumor cells.