Merck
  • Availability and dose response of phytophenols from a wheat bran rich cereal product in healthy human volunteers.
Availability and dose response of phytophenols from a wheat bran rich cereal product in healthy human volunteers.
Molecular nutrition & food research (2016-07-01)
Madalina Neacsu, Jolene McMonagle, Reg J Fletcher, Toine Hulshof, Sylvia H Duncan, Lorraine Scobbie, Gary J Duncan, Louise Cantlay, Graham Horgan, Baukje de Roos, Garry G Duthie, Wendy R Russell
ABSTRACT
Phytophenols present in cereals are metabolised to compounds that could be partly responsible for the reduced risk of chronic diseases and all-cause mortality associated with fibre-rich diets. The bioavailability, form and in vivo concentrations of these metabolites require to be established. Eight healthy volunteers consumed a test meal containing a recommended dose (40 g) and high dose (120 g) of ready-to-eat wheat bran cereal and the systemic and colonic metabolites determined quantitatively by LC-MS. Analysis of the systemic metabolomes demonstrated that a wide range of phytophenols were absorbed/excreted (43 metabolites) within 5 h of consumption. These included 16 of the 21 major parent compounds identified in the intervention product and several of these were also found to be significantly increased in the colon. Not all of the metabolites were increased with the higher dose, suggesting some limitation in absorption due to intrinsic factors and/or the food matrix. Many compounds identified (e.g. ferulic acid and major metabolites) exhibit anti-inflammatory activity and impact on redox pathways. The combination of postprandial absorption and delivery to the colon, as well as hepatic recycling of the metabolites at these concentrations, is likely to be beneficial to both systemic and gut health.
Sigma-Aldrich
Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid