Merck
  • Newly defined conditions for the in vitro cultivation and cryopreservation of Dientamoeba fragilis: new techniques set to fast track molecular studies on this organism.
Newly defined conditions for the in vitro cultivation and cryopreservation of Dientamoeba fragilis: new techniques set to fast track molecular studies on this organism.
Parasitology (2010-07-09)
J L N Barratt, G R Banik, J Harkness, D Marriott, J T Ellis, D Stark
ABSTRACT
Dientamoeba fragilis is a pathogen of the human gastrointestinal tract that is a common cause of diarrhoea. A paucity of knowledge on the in vitro cultivation and cryopreservation of Dientamoeba has meant that few studies have been conducted to investigate its biology. The objective of this study was to define, for the first time, in vitro culture conditions able to support the long-term in vitro growth of Dientamoeba. Also, we aimed to define a suitable method for cryopreserving viable Dientamoeba trophozoites. A modified BD medium, TYGM-9, Loeffler's slope medium, Robinson's medium, Medium 199, Trichosel and a Tritrichomonas fetus medium were compared, using cell counts, for their ability to support the growth of D. fragilis at various temperatures and atmospheric conditions. Loeffler's slope medium supported significantly better growth compared to other media. A temperature of 42°C and a microaerophilic atmosphere were also optimum for Dientamoeba growth. To our knowledge, this is the first study to describe and compare different culture media and conditions for the growth of clinical isolates of D. fragilis. This new technology will aid the development of diagnostics for dientamoebiasis as well as facilitate large-scale sequencing projects that will fast track molecular studies on D. fragilis.