Merck
  • Spatiotemporal dynamics of inner ear sensory and non-sensory cells revealed by single-cell transcriptomics.
Spatiotemporal dynamics of inner ear sensory and non-sensory cells revealed by single-cell transcriptomics.
Cell reports (2021-07-15)
Taha A Jan, Yasmin Eltawil, Angela H Ling, Leon Chen, Daniel C Ellwanger, Stefan Heller, Alan G Cheng
ABSTRACT
The utricle is a vestibular sensory organ that requires mechanosensitive hair cells to detect linear acceleration. In neonatal mice, new hair cells are derived from non-sensory supporting cells, yet cell type diversity and mechanisms of cell addition remain poorly characterized. Here, we perform computational analyses on single-cell transcriptomes to categorize cell types and resolve 14 individual sensory and non-sensory subtypes. Along the periphery of the sensory epithelium, we uncover distinct groups of transitional epithelial cells, marked by Islr, Cnmd, and Enpep expression. By reconstructing de novo trajectories and gene dynamics, we show that as the utricle expands, Islr+ transitional epithelial cells exhibit a dynamic and proliferative phase to generate new supporting cells, followed by coordinated differentiation into hair cells. Taken together, our study reveals a sequential and coordinated process by which non-sensory epithelial cells contribute to growth of the postnatal mouse sensory epithelium.
Sigma-Aldrich
Anti-Sox9 Antibody, Chemicon®, from rabbit
Sigma-Aldrich
Saponin, for molecular biology, used as non-ionic surfactant