Merck
  • MicroRNA-124 facilitates lens epithelial cell apoptosis by inhibiting SPRY2 and MMP-2.
MicroRNA-124 facilitates lens epithelial cell apoptosis by inhibiting SPRY2 and MMP-2.
Molecular medicine reports (2021-03-25)
Yan Liu, Shuting Li, Yao Liu, Xujing Lv, Qing Zhou
ABSTRACT
Age-related cataract (ARC) is the primary cause of blindness worldwide. Abnormal expression of microRNAs (miRNAs/miRs) has been reported to be associated with multiple diseases, including ARC. However, the potential role of miR-124 in ARC remains unclear. The present study used the human lens epithelial cell line, SRA01/04, to investigate the potential role of miR-124 in ARC. Reverse transcription-quantitative PCR analysis was performed to detect the expression levels of miR-124, protein sprouty homolog 2 (SPRY2) and matrix metalloproteinase-2 (MMP-2) in ARC tissues, while western blotting was performed to detect the protein levels of SPRY2 and MMP-2. Cell viability and apoptosis of SRA01/04 cells were assessed via Cell Counting Kit-8 and TUNEL assays, respectively. The interaction between miR-124 and SPRY2 or MMP-2 was confirmed via the dual-luciferase reporter and RNA immunoprecipitation assays. The results of the present study demonstrated that miR-124 expression was significantly upregulated in ARC tissues, and knockdown of miR-124 increased SRA01/04 cell viability and suppressed apoptosis. In addition, SPRY2 and MMP-2 expression was decreased in ARC tissues, and were demonstrated to directly bind to miR-124. Overexpression of SPRY2 or MMP-2 increased SRA01/04 cell viability and repressed apoptosis, the effects of which were reversed following overexpression of miR-124. Taken together, these results suggested that miR-124 facilitates lens epithelial cell apoptosis by modulating SPRY2 or MMP-2 expression, providing a novel treatment approach for ARC.