Merck
  • Nano-liquid chromatography combined with a sustainable microextraction based on natural deep eutectic solvents for analysis of phthalate esters.
Nano-liquid chromatography combined with a sustainable microextraction based on natural deep eutectic solvents for analysis of phthalate esters.
Electrophoresis (2020-04-17)
Álvaro Santana-Mayor, Ruth Rodríguez-Ramos, Bárbara Socas-Rodríguez, Miguel Ángel Rodríguez-Delgado, Giovanni D'Orazio
ABSTRACT
The separation of 11 phthalic acid ester (PAEs) was carried out by nano-liquid chromatography coupled to ultraviolet and MS detection. Preliminary experiments were achieved in order to select suitable stationary phases and chromatographic conditions. The baseline separation was obtained, for all compounds, with an XBridgeTM C18 column in less than 15 min, working in step gradient mode. The sensitivity of the method was improved by on-column focusing. PAEs were extracted from alcoholic and nonalcoholic beverages using vortex-assisted emulsification dispersive liquid-liquid microextration and natural deep eutectic solvents. The whole method was validated in terms of linearity, sensitivity, precision, recovery, and repeatability. Combination of both off-line sample preparation preconcentration and large injection volume led to obtain LOQs in the range 5-47 ng/mL. The developed nano-LC-UV method was extended to MS detection to confirm the presence of PAEs in some beverages commercialized in different types of packaging.