Merck
  • Helmet Phthalocyaninato Iron Complex as a Primary Drier for Alkyd Paints.
Helmet Phthalocyaninato Iron Complex as a Primary Drier for Alkyd Paints.
Materials (Basel, Switzerland) (2021-04-04)
Jan Honzíček, Eliška Matušková, Štěpán Voneš, Jaromír Vinklárek
ABSTRACT
This study describes the catalytic performance of an iron(III) complex bearing a phthalocyaninato-like ligand in two solvent-borne and two high-solid alkyd binders. Standardized mechanical tests revealed strong activity, which appeared in particular cases at concentrations about one order of magnitude lower than in the case of cobalt(II) 2-ethylhexanoate, widespread used in paint-producing industry. The effect of the iron(III) compound on autoxidation process, responsible for alkyd curing, was quantified by kinetic measurements by time-resolved infrared spectroscopy and compared with several primary driers. Effect of the drier concentration on coloration of transparent coatings was determined by UV-Vis spectroscopy.