Merck
  • Structural Reorganization of Imidazolium Ionic Liquids Induced by Pressure-Enhanced Ionic Liquid-Polyethylene Oxide Interactions.
Structural Reorganization of Imidazolium Ionic Liquids Induced by Pressure-Enhanced Ionic Liquid-Polyethylene Oxide Interactions.
International journal of molecular sciences (2021-01-23)
Teng-Hui Wang, Li-Wen Hsu, Hai-Chou Chang
ABSTRACT
Mixtures of polyethylene oxide (PEO, M.W.~900,000) and imidazolium ionic liquids (ILs) are studied using high-pressure Fourier-transform infrared spectroscopy. At ambient pressure, the spectral features in the C-H stretching region reveal that PEO can disturb the local structures of the imidazolium rings of [BMIM]+ and [HMIM]+. The pressure-induced phase transition of pure 1-butyl-3-methylimidazolium bromide ([BMIM]Br) is observed at a pressure of 0.4 GPa. Pressure-enhanced [BMIM]Br-PEO interactions may assist PEO in dividing [BMIM]Br clusters to hinder the aggregation of [BMIM]Br under high pressures. The C-H absorptions of pure 1-hexyl-3-methylimidazolium bromide [HMIM]Br do not show band narrowing under high pressures, as observed for pure [BMIM]Br. The band narrowing of C-H peaks is observed at 1.5 GPa for the [HMIM]Br-PEO mixture containing 80 wt% of [HMIM]Br. The presence of PEO may reorganize [HMIM]Br clusters into a semi-crystalline network under high pressures. The differences in aggregation states for ambient-pressure phase and high-pressure phase may suggest the potential of [HMIM]Br-PEO (M.W.~900,000) for serving as optical or electronic switches.