Merck
  • HEMA-induced oxidative stress inhibits NF-κB nuclear translocation and TNF release from LTA- and LPS-stimulated immunocompetent cells.
HEMA-induced oxidative stress inhibits NF-κB nuclear translocation and TNF release from LTA- and LPS-stimulated immunocompetent cells.
Dental materials : official publication of the Academy of Dental Materials (2020-12-12)
Helmut Schweikl, Margaritha Birke, Marialucia Gallorini, Christine Petzel, Carola Bolay, Claudia Waha, Karl-Anton Hiller, Wolfgang Buchalla
ABSTRACT
The release of inflammatory cytokines from antigen-stimulated cells of the immune system is inhibited by resin monomers such as 2-hydroxyethyl methacrylate (HEMA). Although the formation of oxidative stress in cells exposed to HEMA is firmly established, the mechanism behind the inhibited cytokine secretion is only partly known. The present investigation presents evidence regarding the role of HEMA-induced oxidative stress in the secretion of the pro-inflammatory cytokine TNFα from cells exposed to the antigens LTA (lipoteichoic acid) or LPS (lipopolysaccharide) of cariogenic microorganisms using BSO (L-buthionine sulfoximine) or NAC (N-acetyl cysteine) to inhibit or stabilize the amounts of the antioxidant glutathione. RAW264.7 mouse macrophages were treated with LTA, LPS or HEMA in the presence of BSO or NAC for 1h or 24h. Secretion of TNFα from cell cultures was analyzed by ELISA, and the formation of reactive oxygen (ROS) or nitrogen species (RNS) was determined by flow cytometry. Protein expression was detected by Western blotting. The release of TNFα in both LTA- and LPS-exposed cells was decreased by HEMA, and this concentration-dependent inhibitory effect was amplified by BSO or NAC. LTA- and LPS-stimulated expression of the redox-sensitive transcription factor NF-αB (p65) in cell nuclei decreased in the presence of HEMA because the translocation of p65 from the cytosol was prevented by oxidative stress specifically increased by the monomer. A disturbance of the cellular redox balance, particularly induced by HEMA, is a crucial factor in the inhibition of LTA- and LPS-stimulated signalling pathways leading to TNFα secretion.