Merck
  • PolyMet-HA nanocomplexs regulates glucose uptake by inhibiting SHIP2 activity.
PolyMet-HA nanocomplexs regulates glucose uptake by inhibiting SHIP2 activity.
Journal of biomaterials applications (2020-08-04)
Xinlu Yuan, Ling Ding, Jianjun Diao, Song Wen, Chenglin Xu, Ligang Zhou, Anqing Du
ABSTRACT
Metformin, the first-line drug to treat type 2 diabetes, inhibits mitochondrial glycerolphosphate dehydrogenase in the liver to suppress gluconeogenesis. The major adverse effects caused by metformin were lactic acidosis and gastrointestinal discomfort. Therefore, there is need to develop a strategy with excellent permeability and appropriate retention effects.In this study, we synthesized a simple and biocompatible PolyMetformin (denoted as PolyMet) through conjugation of PEI1.8K with dicyandiamide, and then formed PolyMet-hyaluronic acid (HA) nanocomplexs by electrostatic self-assembly of the polycationic PolyMet and polyanionic hyaluronic acid (HA). Similar to metformin, the PolyMet-HA nanocomplexs could reduce the catalytic activity of the recombinant SHIP2 phosphatase domain in vitro. In SHIP2-overexpressing myotubes, PolyMet-HA nanocomplexes ameliorated glucose uptake by downregulating glucose transporter 4 endocytosis. PolyMet-HA nanocomplexes also could restore Akt signaling and protect the podocyte from apoptosis induced by SHIP2 overexpression. In essence, the PolyMet-HA nanocomplexes act similarly to metformin and increase glucose uptake, and maybe have a potential role in the treatment of type 2 diabetes.