Merck
  • Upregulating microRNA-874-3p inhibits CXCL12 expression to promote angiogenesis and suppress inflammatory response in ischemic stroke.
Upregulating microRNA-874-3p inhibits CXCL12 expression to promote angiogenesis and suppress inflammatory response in ischemic stroke.
American journal of physiology. Cell physiology (2020-07-02)
Kangling Xie, Ying Cai, Pu Yang, Feng Du, Kai Wu
ABSTRACT
Identification of specific biomarkers for ischemic stroke is necessary due to their abilities to improve treatment outcomes. Many studies have demonstrated the involvement of microRNAs (miRNAs) in the pathogenesis and complications of ischemic stroke and patient outcomes. We found that the expression of miR-874-3p was downregulated in clinical samples of ischemic stroke. Thus the present study explored the potential role of miR-874-3p in ischemic stroke and related mechanisms. A mouse model of ischemic stroke was constructed by middle cerebral artery occlusion. The relationship among miR-874-3p, C-X-C motif chemokine ligand 12 (CXCL12), and the Wnt/β-catenin pathway was explored by dual luciferase reporter assay and Western blot analysis. Angiogenesis and brain tissue apoptosis were evaluated by immunofluorescence staining and TUNEL staining, respectively. ELISA was introduced to measure levels of inflammatory factors, including tumor necrosis factor-α (TNF-α), interleukin (IL)-1, IL-6, IL-8, and IL-10 in brain tissues. Primary hippocampal neuronal cells were isolated from the mouse model of ischemic stroke and incubated with human umbilical vein endothelial cells (HUVECs) for HUVEC tube formation. High expression of CXCL12 and low expression of miR-874-3p were confirmed in ischemic stroke. In addition, miR-874-3p was found to target and downregulate CXCL12, thus reducing TNF-α, IL-1, IL-6, and IL-8 levels, but enhancing IL-10 level. Collectively, upregulating miR-874-3p inhibits CXCL12 expression to promote angiogenesis and inhibit inflammation in ischemic stroke mice by activating the Wnt/β-catenin pathway, which may provide a new direction of ischemic stroke treatment.