Merck
  • Circ-XPR1 promotes osteosarcoma proliferation through regulating the miR-214-5p/DDX5 axis.
Circ-XPR1 promotes osteosarcoma proliferation through regulating the miR-214-5p/DDX5 axis.
Human cell (2020-09-14)
Xiaohuan Mao, Shuren Guo, Lan Gao, Gang Li
ABSTRACT
Circular RNAs (circRNAs) are a new class of RNAs that play an important role in the development of various tumors. However, the expression profile and biological function of circRNAs in osteosarcoma (OS) progression remain unclear. OS-related circRNA expression profiles from the GEO database (GSE96964) were downloaded to identify differentially expressed circRNAs between OS and normal tissues. We identified one upregulated circRNA (Circ-XPR1), and RT-PCR was performed to further confirm the expression abundance in OS tissue. Circ-XPR1 was closely related to overall survival and disease-free survival of OS patients. Knockdown of Circ-XPR1 significantly reduced the proliferation of OS cells. Gain- and loss-of-function studies showed that Circ-XPR1 promoted OS cell proliferation by sponging miR-214-5p to regulate DDX5 expression. Our findings suggested that Circ-XPR1 regulates OS cell proliferation by sponging miR-214-5p to regulate DDX5 expression. Therefore, the Circ-XPR1/miR-214-5p/DDX5 axis may serve as a potential therapeutically relevant target for OS.