Merck
  • Enhancement versus neutralization by SARS-CoV-2 antibodies from a convalescent donor associates with distinct epitopes on the RBD.
Enhancement versus neutralization by SARS-CoV-2 antibodies from a convalescent donor associates with distinct epitopes on the RBD.
Cell reports (2021-01-25)
Yunjiao Zhou, Zezhong Liu, Shibo Li, Wei Xu, Qianqian Zhang, Israel T Silva, Cheng Li, Yanling Wu, Qingling Jiang, Zhenmi Liu, Qiujing Wang, Yu Guo, Jianbo Wu, Chengjian Gu, Xia Cai, Di Qu, Christian T Mayer, Xiangxi Wang, Shibo Jiang, Tianlei Ying, Zhenghong Yuan, Youhua Xie, Yumei Wen, Lu Lu, Qiao Wang
ABSTRACT
Several potent neutralizing antibodies against severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) virus have been identified. However, antibody-dependent enhancement (ADE) has not been comprehensively studied for SARS-CoV-2, and the relationship between enhancing versus neutralizing activities and antibody epitopes remains unknown. Here, we select a convalescent individual with potent IgG neutralizing activity and characterize his antibody response. Monoclonal antibodies isolated from memory B cells target four groups of five non-overlapping receptor-binding domain (RBD) epitopes. Antibodies to one group of these RBD epitopes mediate ADE of entry in Raji cells via an Fcγ receptor-dependent mechanism. In contrast, antibodies targeting two other distinct epitope groups neutralize SARS-CoV-2 without ADE, while antibodies against the fourth epitope group are poorly neutralizing. One antibody, XG014, potently cross-neutralizes SARS-CoV-2 variants, as well as SARS-CoV-1, with respective IC50 (50% inhibitory concentration) values as low as 5.1 and 23.7 ng/mL, while not exhibiting ADE. Therefore, neutralization and ADE of human SARS-CoV-2 antibodies correlate with non-overlapping RBD epitopes.
Sigma-Aldrich
IGEPAL® CA-630, for molecular biology