Merck
  • ΔMST and the Regulation of Cardiac CSE and OTR Expression in Trauma and Hemorrhage.
ΔMST and the Regulation of Cardiac CSE and OTR Expression in Trauma and Hemorrhage.
Antioxidants (Basel, Switzerland) (2021-02-07)
Britta Trautwein, Tamara Merz, Nicole Denoix, Csaba Szabo, Enrico Calzia, Peter Radermacher, Oscar McCook
ABSTRACT
Genetic deletion of 3-mercaptopyruvate sulfurtransferase (MST) is known to result in hypertension and cardiac hypertrophy in older mice, and is associated with increased anxiety-like behaviors. Endogenous hydrogen sulfide (H2S) produced by MST in the mitochondria is also known to be involved in physiological and cellular bioenergetics, and its dysfunction associated with depressive behavior and increased cardiovascular morbidity. Interestingly, early life stress has been shown to lead to a significant loss of cystathionine-γ-lyase (CSE) and oxytocin receptor (OTR) expression in the heart. Thus, we were interested in testing the hypothesis of whether genetic MST mutation (ΔMST) would affect cardiac CSE and OTR expression and affect the mitochondrial respiration in a clinically relevant, resuscitated, mouse model of trauma and hemorrhagic shock. In ΔMST mice, we found a reduction of CSE and OTR in both the naive as well as injured state, in contrast to the wild type (wt) controls. Interestingly, the ΔMST showed a different complex IV response to injury than the wt controls, although our claims are based on the non-demonstrated assumption that naive wt and naive ΔMST mice have comparable complex IV activity. Finally, hemorrhagic shock led to a reduction of CSE and OTR, confirming previous results in the injured mouse heart. To date, the exact mechanisms of the cardiac interaction between H2S and OT are not clear, but they point the way to potential cardioprotective therapies.