Merck
  • MS Binding Assays for Glycine Transporter 2 (GlyT2) Employing Org25543 as Reporter Ligand.
MS Binding Assays for Glycine Transporter 2 (GlyT2) Employing Org25543 as Reporter Ligand.
ChemMedChem (2020-08-01)
Thomas M Ackermann, Lars Allmendinger, Georg Höfner, Klaus T Wanner
ABSTRACT
This study describes the first binding assay for glycine transporter 2 (GlyT2) following the concept of MS Binding Assays. The selective GlyT2 inhibitor Org25543 was employed as a reporter ligand and it was quantified with a highly sensitive and rapid LC-ESI-MS/MS method. Binding of Org25543 at GlyT2 was characterized in kinetic and saturation experiments with an off-rate of 7.07×10-3  s-1 , an on-rate of 1.01×106  M-1  s-1 , and an equilibrium dissociation constant of 7.45 nM. Furthermore, the inhibitory constants of 19 GlyT ligands were determined in competition experiments. The validity of the GlyT2 affinities determined with the binding assay was examined by a comparison with published inhibitory potencies from various functional assays. With the capability for affinity determination towards GlyT2 the developed MS Binding Assays provide the first tool for affinity profiling of potential ligands and it represents a valuable new alternative to functional assays addressing GlyT2.