Merck
  • Identification of the bacteria and their metabolic activities associated with the microbial spoilage of custard cream desserts.
Identification of the bacteria and their metabolic activities associated with the microbial spoilage of custard cream desserts.
Food microbiology (2019-11-11)
Clarisse Techer, Sophie Jan, Anne Thierry, Marie-Bernadette Maillard, Noël Grosset, Olivier Galet, Valérie Breton, Michel Gautier, Florence Baron
ABSTRACT
The famous French dessert "ile flottante" consists of a sweet egg white foam floating on a vanilla custard cream, which contains highly nutritive raw materials, including milk, sugar and egg. Spoilage issues are therefore a key concern for the manufacturers. This study explored the bacterial diversity of 64 spoiled custard cream desserts manufactured by 2 French companies. B. cereus group bacteria, coagulase negative Staphylococcus, Enterococcus and Leuconostoc spp. were isolated from spoiled products. Thirty-one bacterial isolates representative of the main spoilage species were tested for their spoilage abilities. Significant growth and pH decrease were observed regardless of species. While off-odours were detected with B. cereus group and staphylococci, yoghurt odours were detected with Enterococcus spp. and Leuconostoc spp. B. cereus group bacteria produced various esters and several compounds derived from amino acid and sugar metabolism. Most Staphylococci produced phenolic compounds. Enterococcus spp. and Leuconostoc spp. isolates produced high levels of compounds derived from sugar metabolism. Each type of spoilage bacteria was associated with a specific volatile profile and lactic acid was identified as a potential marker of spoilage of custard cream-based desserts. These findings provide valuable information for manufacturers to improve food spoilage detection and prevention of chilled desserts made with milk and egg.