Merck
  • A dual-acting 5-HT6 receptor inverse agonist/MAO-B inhibitor displays glioprotective and pro-cognitive properties.
A dual-acting 5-HT6 receptor inverse agonist/MAO-B inhibitor displays glioprotective and pro-cognitive properties.
European journal of medicinal chemistry (2020-09-20)
Vittorio Canale, Katarzyna Grychowska, Rafał Kurczab, Mateusz Ryng, Abdul Raheem Keeri, Grzegorz Satała, Agnieszka Olejarz-Maciej, Paulina Koczurkiewicz, Marcin Drop, Klaudia Blicharz, Kamil Piska, Elżbieta Pękala, Paulina Janiszewska, Martyna Krawczyk, Maria Walczak, Severine Chaumont-Dubel, Andrzej J Bojarski, Philippe Marin, Piotr Popik, Paweł Zajdel
ABSTRACT
The complex etiology of Alzheimer's disease has initiated a quest for multi-target ligands to address the multifactorial causes of this neurodegenerative disorder. In this context, we designed dual-acting 5-HT6 receptor (5-HT6R) antagonists/MAO-B inhibitors using pharmacophore hybridization strategy. Our approach involved linking priviliged scaffolds of 5-HT6R with aryloxy fragments derived from reversible and irreversible MAO-B inhibitors. The study identified compound 48 that acts as an inverse agonist of 5-HT6R at Gs signaling and an irreversible MAO-B inhibitor. Compound 48 showed moderate metabolic stability in rat microsomal assay, artificial membrane permeability, no hepatotoxicity, and it was well distributed to the brain. Additionally, 48 showed glioprotective properties in a model of cultured astrocytes using 6-OHDA as the cytotoxic agent. Finally, compound 48 (MED = 1 mg/kg, p.o.) fully reversed memory deficits in the NOR task induced by scopolamine in rats. A better understanding of effects exerted by dual-acting 5-HT6R/MAO-B modulators may impact the future development of neurodegenerative-directed treatment strategies.