- Heterozygous Ile453Val codon mutation in exon 7, homozygous single nucleotide polymorphisms in intron 2 and 5 of cathepsin C are associated with Haim-Munk syndrome.
Heterozygous Ile453Val codon mutation in exon 7, homozygous single nucleotide polymorphisms in intron 2 and 5 of cathepsin C are associated with Haim-Munk syndrome.
European journal of dentistry (2014-06-27)
Nalini Aswath, Bhuminathan Swamikannu, Sankar Narayanan Ramakrishnan, Rajendran Shanmugam, Jayakar Thomas, Arvind Ramanathan
PMID24966751
ABSTRACT
In the present study, we have investigated the genetic status of CTSC gene in a HMS subject, who along with her parents belonged to non-Jewish South Indian Dravidian community. Genomic deoxyribonucleic acid isolated from the peripheral blood of the subject was amplified with CTSC exon specific primers and were analyzed by direct sequencing. Sequencing analysis identified Ile453Val mutation within exon 7 of CTSC gene in heterozygous condition, and two single nucleotide polymorphisms (SNPs) within intron 2 and 5 in homozygous condition. The present study has identified for the first time the association of Ile453Val mutation within exon 7 and the two SNPs in a subject with HMS.