Merck
  • 18F-Fluciclovine PET metabolic imaging reveals prostate cancer tumour heterogeneity associated with disease resistance to androgen deprivation therapy.
18F-Fluciclovine PET metabolic imaging reveals prostate cancer tumour heterogeneity associated with disease resistance to androgen deprivation therapy.
EJNMMI research (2020-11-26)
Gaurav Malviya, Rachana Patel, Mark Salji, Rafael S Martinez, Peter Repiscak, Ernest Mui, Susan Champion, Agata Mrowinska, Emma Johnson, Maha AlRasheedi, Sally Pimlott, David Lewis, Hing Y Leung
ABSTRACT
Prostate cancer is highly prevalent worldwide. Androgen deprivation therapy (ADT) remains the treatment of choice for incurable prostate cancer, but majority of patients develop disease recurrence following ADT. There is therefore an urgent need for early detection of treatment resistance. Isogenic androgen-responsive (CWR22Res) and castration-resistant (22Rv1) human prostate cancer cells were implanted into the anterior lobes of the prostate in CD-1 Nu mice to generate prostate orthografts. Castrated mice bearing CWR22Res and 22Rv1 orthografts mimic clinical prostate cancer following acute and chronic ADT, respectively. 18F-Fluciclovine (1-amino-3-fluorocyclobutane-1-carboxylic acid) with a radiochemical purity of > 99% was produced on a FASTlab synthesiser. Ki67 staining in endpoint orthografts was studied. Western blot, quantitative RT-PCR and next-generation sequencing transcriptomic analyses were performed to assess the expression levels of amino acid transporters (including LAT1 and ASCT2, which have been implicated for Fluciclovine uptake). Longitudinal metabolic imaging with 18F-Fluciclovine-based positron emission tomography (PET) was performed to study tumour response following acute and chronic ADT. Both immunohistochemistry analysis of endpoint prostate tumours and longitudinal 18F-Fluciclovine imaging revealed tumour heterogeneity, particularly following ADT, with in vivo 18F-Fluciclovine uptake correlating to viable cancer cells in both androgen-proficient and castrated environment. Highlighting tumour subpopulation following ADT, both SUVpeak and coefficient of variation (CoV) values of 18F-Fluciclovine uptake are consistent with tumour heterogeneity revealed by immunohistochemistry. We studied the expression of amino acid transporters (AATs) for 18F-Fluciclovine, namely LAT1 (SLC7A5 and SLC3A2) and ASCT2 (SLC1A5). SLC7A5 and SLC3A2 were expressed at relatively high levels in 22Rv1 castration-resistant orthografts following chronic ADT (modelling clinical castration-resistant disease), while SLC1A5 was preferentially expression in CWR22Res tumours following acute ADT. Additional AATs such as SLC43A2 (LAT4) were shown to be upregulated following chronic ADT by transcriptomic analysis; their role in Fluciclovine uptake warrants investigation. We studied in vivo 18F-Fluciclovine uptake in human prostate cancer orthograft models following acute and chronic ADT. 18F-Fluciclovine uptakes highlight tumour heterogeneity that may explain castration resistance and can be exploited as a clinical biomarker.