Merck
  • Investigation of ruminant xenobiotic metabolism in a modified rumen simulation system (RUSITEC).
Investigation of ruminant xenobiotic metabolism in a modified rumen simulation system (RUSITEC).
ALTEX (2018-03-15)
Barbara Birk, Alexander Stähle, Mathias Meier, Markus Palm, Dorothee Funk-Weyer, Gerhard Breves, Harald Seulberger
ABSTRACT
The approving agencies for plant protection agents request xenobiotic metabolism and residue studies in rats, farm animals and plants (e.g. EU regulation 1107/2009) according to OECD guidelines. The specific intestinal physiology of ruminants might lead to specific residues, which should be investigated very carefully. Specific aspects of xenobiotic metabolism in ruminants may arise, which are investigated by performing additional in vivo studies. The aim of the present work is to asses a modified rumen simulation system (RUSITEC) for such studies in vitro. Rumen constituents from sheep were incubated over 8 days. Physiological parameters followed by monitoring the pH (6.70 ± 0.07), the redox potential (301 mV ± 30 mV), the microbial composition and determination of β-glucosidase activity. After anequilibration period of three to four days the fermenters were probed with 14C-labelled triazolederivatives, i.e.common metabolites of azole fungicides. Only triazole-alanine was cleaved to 1,2,4-triazole, while triazole-acetic acid and triazole-lactic acid remained stable up to 96 h. Moreover, glycosides are often determined as the main residues in the plants. This analysis showed, that the two glucosides octyl-β-D-glucopyranoside and polydatin were both rapidly cleaved in this rumen in vitro system. These data showed that the modified RUSITEC system is stable, viable and maintained metabolic capacity over a longer period (at least 8 days). This makes many animal experiments obsolete and lead to a significant contribution of the 3R (refine, reduce, replace). The modification of the RUSITEC system enables safe routine use for unlabeled but also for radioactive labelled compounds.