Merck
  • Gonadotropin-regulated testicular helicase (DDX25), an essential regulator of spermatogenesis, prevents testicular germ cell apoptosis.
Gonadotropin-regulated testicular helicase (DDX25), an essential regulator of spermatogenesis, prevents testicular germ cell apoptosis.
The Journal of biological chemistry (2008-04-24)
Ravi K Gutti, Chon-Hwa Tsai-Morris, Maria L Dufau
ABSTRACT
Gonadotropin-regulated testicular helicase (GRTH)/DDX25 is an essential post-transcriptional regulator of spermatogenesis. In GRTH null mice severe apoptosis was observed in spermatocytes entering the metaphase of meiosis. Pro- and anti-apoptotic factors were found to be under GRTH regulation in comparative studies of spermatocytes from wild type and GRTH(-/-) knock-out (KO) mice. KO mice displayed decreased levels of Bcl-2 and Bcl-xL (anti-apoptotic factors), an increase in Bid, Bak, and Bad (pro-apoptotic), reduced phospho-Bad, and release of cytochrome c. Also, an increase on Smac, a competitor of inhibitor apoptotic proteins that release caspases, was observed. These changes caused an increase in cleavage of caspases 9 and 3, activation of caspase 3 and increases in cleavage products of PARP. The half-life of caspase 3 transcripts was markedly increased in KO, indicating that GRTH had a negative role on its mRNA stability. IkappaBalpha, which sequesters NF-kappaB from its transcriptional activation of pro-apoptotic genes, was highly elevated in KO, and its phospho-form, which promotes its dissociation, was reduced. The increase of HDAC1 and abolition of p300 expression in KO indicated a nuclear action of GRTH on the NF-kappaB-mediated transcription of anti-apoptotic genes. It also regulates the associated death domain pathway and caspase 8-mediated events. GRTH-mediated apoptotic regulation was further indicated by its selective binding to pro- and anti-apoptotic mRNAs. These studies have demonstrated that GRTH, as a component of mRNP particles, acts as a negative regulator of the tumor necrosis factor receptor 1 and caspase pathways and promotes NF-kappaB function to control apoptosis in spermatocytes of adult mice.