Merck
  • vacA Genotype Status of Helicobacter pylori Isolated from Foods with Animal Origin.
vacA Genotype Status of Helicobacter pylori Isolated from Foods with Animal Origin.
BioMed research international (2016-04-19)
Elnaz Saeidi, Amirhossein Sheikhshahrokh
ABSTRACT
According to controversial theories and results of studies, foods with animal origins play an important role in the transmission of H. pylori to human. The aim of this study was to determine the distribution of vacA genotypes of H. pylori, isolated from milk and meat samples of cow, sheep, goat, camel, and buffalo. Eight hundred and twenty raw milk and meat samples were collected from various parts of Iran. Samples were cultured and those found positive for H. pylori were analyzed for the presence of various genotypes of vacA gene. Out of 420 milk and 400 meat samples, 92 (21.90%) and 105 (26.25%) were positive for H. pylori, respectively. The most commonly detected genotypes in the vacA gene were s1a (86.80%), m1a (79.18%), s1b (69.54%), and m1b (63.45%) and detected combined genotypes were mostly m1as1a (68.52%), m1as1b (60.40%), m1bs1b (55.83%), and m1bs1a (53.29%). High presence of bacteria in the milk and meat samples of sheep represents that sheep may be the natural host of H. pylori. High presence of H. pylori strains in milk and meat samples similar to vacA genotypes in human being suggests that milk and meat samples could be the sources of bacteria for human.