Merck
  • Comparative Chromosome Painting and NOR Distribution Suggest a Complex Hybrid Origin of Triploid Lepidodactylus lugubris (Gekkonidae).
Comparative Chromosome Painting and NOR Distribution Suggest a Complex Hybrid Origin of Triploid Lepidodactylus lugubris (Gekkonidae).
PloS one (2015-07-07)
Vladimir A Trifonov, Alessio Paoletti, Vincenzo Caputo Barucchi, Tatiana Kalinina, Patricia C M O'Brien, Malcolm A Ferguson-Smith, Massimo Giovannotti
ABSTRACT
Parthenogenesis, unisexuality and triploidy are interesting but poorly studied phenomena occurring in some reptile species. The mourning gecko (Lepidodactylus lugubris) represents a complex of diploid and triploid parthenogenetic mostly all-female populations (males occur quite rarely) widely distributed in coastal areas of the Indian and Pacific Oceans. Here, we study karyotypes of a male and two female L. lugubris (LLU) triploid individuals (3n = 66) using comparative painting with Gekko japonicus, Hemidactylus turcicus and H. platyurus chromosome specific probes to visualize the homologous regions and to reveal genus specific rearrangements. Also, we applied a 28S ribosomal DNA probe and Ag-staining to detect nucleolus organizer regions (NORs). Our results suggest that the karyotype of L. lugubris underwent a chromosome fission and a fusion after its divergence from a common ancestor of the Gekko-Hemidactylus group. The NORs were found to be located on one out of three homologs on each of LLU8, LLU15 and LLU18, thus further confirming a hybrid origin of triploid individuals. It seems that three different bisexual populations might have contributed to the origin of this triploid parthenogenetic population. We postulate that the heterozygosity in NOR localization is maintained in the triploid clone studied by the absence of recombination as described in whiptail lizards. The pattern of NOR localizations and homologous regions in males and females, as well as the absence of other detectable karyotypic differences, suggest that males arise spontaneously in all female populations and do not arise from independent hybridizations with different species.