Merck
  • Effects of Eyjafjallajökull volcanic ash on innate immune system responses and bacterial growth in vitro.
Effects of Eyjafjallajökull volcanic ash on innate immune system responses and bacterial growth in vitro.
Environmental health perspectives (2013-03-13)
Martha M Monick, Jonas Baltrusaitis, Linda S Powers, Jennifer A Borcherding, Juan C Caraballo, Imali Mudunkotuwa, David W Peate, Katherine Walters, Jay M Thompson, Vicki H Grassian, Gunnar Gudmundsson, Alejandro P Comellas
ABSTRACT
On 20 March 2010, the Icelandic volcano Eyjafjallajökull erupted for the first time in 190 years. Despite many epidemiological reports showing effects of volcanic ash on the respiratory system, there are limited data evaluating cellular mechanisms involved in the response to ash. Epidemiological studies have observed an increase in respiratory infections in subjects and populations exposed to volcanic eruptions. We physicochemically characterized volcanic ash, finding various sizes of particles, as well as the presence of several transition metals, including iron. We examined the effect of Eyjafjallajökull ash on primary rat alveolar epithelial cells and human airway epithelial cells (20-100 µg/cm(2)), primary rat and human alveolar macrophages (5-20 µg/cm(2)), and Pseudomonas aeruginosa (PAO1) growth (3 µg/104 bacteria). Volcanic ash had minimal effect on alveolar and airway epithelial cell integrity. In alveolar macrophages, volcanic ash disrupted pathogen-killing and inflammatory responses. In in vitro bacterial growth models, volcanic ash increased bacterial replication and decreased bacterial killing by antimicrobial peptides. These results provide potential biological plausibility for epidemiological data that show an association between air pollution exposure and the development of respiratory infections. These data suggest that volcanic ash exposure, while not seriously compromising lung cell function, may be able to impair innate immunity responses in exposed individuals.