Merck
Targeting tumor cells based on Phosphodiesterase 3A expression.
Experimental cell research (2017-11-07)
Madiha Nazir, Wojciech Senkowski, Frida Nyberg, Kristin Blom, Per-Henrik Edqvist, Malin Jarvius, Claes Andersson, Mats G Gustafsson, Peter Nygren, Rolf Larsson, Mårten Fryknäs
ABSTRACT
We and others have previously reported a correlation between high phosphodiesterase 3A (PDE3A) expression and selective sensitivity to phosphodiesterase (PDE) inhibitors. This indicates that PDE3A could serve both as a drug target and a biomarker of sensitivity to PDE3 inhibition. In this report, we explored publicly available mRNA gene expression data to identify cell lines with different PDE3A expression. Cell lines with high PDE3A expression showed marked in vitro sensitivity to PDE inhibitors zardaverine and quazinone, when compared with those having low PDE3A expression. Immunofluorescence and immunohistochemical stainings were in agreement with PDE3A mRNA expression, providing suitable alternatives for biomarker analysis of clinical tissue specimens. Moreover, we here demonstrate that tumor cells from patients with ovarian carcinoma show great variability in PDE3A protein expression and that level of PDE3A expression is correlated with sensitivity to PDE inhibition. Finally, we demonstrate that PDE3A is highly expressed in subsets of patient tumor cell samples from different solid cancer diagnoses and expressed at exceptional levels in gastrointestinal stromal tumor (GIST) specimens. Importantly, vulnerability to PDE3 inhibitors has recently been associated with co-expression of PDE3A and Schlafen family member 12 (SLFN12). We here demonstrate that high expression of PDE3A in clinical specimens, at least on the mRNA level, seems to be frequently associated with high SLFN12 expression. In conclusion, PDE3A seems to be both a promising biomarker and drug target for individualized drug treatment of various cancers.