Merck
  • Purification and characterization of a newly serine protease inhibitor from Rhamnus frangula with potential for use as therapeutic drug.
Purification and characterization of a newly serine protease inhibitor from Rhamnus frangula with potential for use as therapeutic drug.
3 Biotech (2017-06-10)
Abir Ben Bacha, Ikram Jemel, Nadine M S Moubayed, Imen Ben Abdelmalek
ABSTRACT
Protease inhibitors from plants are well known to be potent inhibitors of the growth of bacteria, fungi, and even certain viruses which make them excellent candidates for use as the lead compounds for the development of novel antimicrobial agents for applications in medicine. In this study, Rhamnus frangula was selected as a protease inhibitor source. The maximum recovery of the protease inhibitor against trypsin was recorded in the crude extract made in 0.1 M phosphate buffer (pH 7.0) and isolated from the mature leaves. Then, the protease inhibitor designated as RfIP1 was purified to homogeneity by Sephadex G50 with an apparent molecular mass of 22.5 kDa and its N-terminal sequence exhibited a high degree of homology with known serine protease inhibitor sequences. The RfIP1 displayed maximal activity at pH 7 and 37 °C. It maintained almost 80% of its maximal activity through a large pH range. The thermo-stability of RfIP1 was markedly enhanced by BSA, CaCl2, and sorbitol, whereas the addition of Mg2+, Zn2+, NaTDC, SDS, DTT, and β-ME significantly promoted inhibitory activity. The protease inhibitor displayed high inhibitory activity toward some known proteases (cathepsin B, chymotrypsin, collagenase, thrombin, and trypsin) that have more importance in pharmaceutical industry and it acted as potent inhibitor of some commercially proteases from Aspergillus oryzae, Bacillus sp, and Bacillus licheniformis. The protease inhibitor also possessed an appreciable antibacterial effect against both Gram-positive and Gram-negative bacteria.