Merck
  • One-step production of α-ketoglutaric acid from glutamic acid with an engineered L-amino acid deaminase from Proteus mirabilis.
One-step production of α-ketoglutaric acid from glutamic acid with an engineered L-amino acid deaminase from Proteus mirabilis.
Journal of biotechnology (2013-01-22)
Long Liu, Gazi Sakir Hossain, Hyun-dong Shin, Jianghua Li, Guocheng Du, Jian Chen
ABSTRACT
Currently, α-ketoglutaric acid (α-KG) is industrially produced by multi-step chemical synthesis, which can cause heavy environmental pollution. Here we reported a simple one-step approach for the production of α-KG by transforming l-glutamic acid with an engineered l-amino acid deaminase (l-AAD) from Proteus mirabilis. First, to facilitate the purification of membrane-bound l-AAD, one N-terminal transmembrane region (from 21 to 87th nucleotide) was removed from l-AAD to block the binding of l-AAD with membrane, and the relatively low-usage codons were replaced by high-usage codons in Escherichia coli to improve the expression level. However, inclusion bodies formed when expressing the ΔN-LAAD in E. coli BL 21, and then the soluble and active ΔN-LAAD was obtained by the solubilization and renaturation of ΔN-LAAD. Furthermore, the biochemical properties of the refolded ΔN-LAAD were characterized and compared with those of full-length l-AAD. Finally, the ΔN-LAAD was used to synthesize α-KG and the maximal formation rate of α-KG reached 12.6% (w/w) at 6h under the following conditions: 12g/L l-glutamic acid, 0.1g/L ΔN-LAAD, 5mM MgCl2, temperature 45°C and pH 8.0. Compared with the multi-step chemical synthesis, the transformation approach has less environmental pollution and has a great potential for α-KG production.